Sodium fluoride ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1141-100G
93-1141-500G
100204-146EA
65.11
USD
100204-146
100211-202
Sodium fluoride ≥99% ACS
Sodium fluoride
Formula:
NaF Boiling Pt: 1695 °C Melting Pt: 988 °C Density: 2.558 Storage Temperature: Ambient |
MDL Number:
MFCD00003524 CAS Number: 7681-49-4 EINECS: 231-667-8 UN: 1690 ADR: 6.1,III Merck Index: 13,08691 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...