Sodium chloride ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Saline, NaCl, Sea salt, Halite
93-1128-1000G
93-1128-5000G
100208-494EA
55.2
USD
100208-494
100210-158
Sodium chloride ≥99% ACS
Sodium chloride
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 Melting Pt: 801 Density: 2.165 Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...