Sodium bromide ≥99.999% (Na basis), PURATREM

Supplier: STREM CHEMICALS INC MS

93-1121-50G 93-1121-10G
100201-858EA 386.4 USD
100201-858 100204-138
Sodium bromide ≥99.999% (Na basis), PURATREM
Sodium bromide
Formula: NaBr
Boiling Pt: 1390 °C
Melting Pt: 755 °C
Density: 3.203
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR