Sodium aluminate ≥99.9% (Al basis) contains ~8% H2O

Supplier: STREM CHEMICALS INC MS
Danger

93-1108-2000G 93-1108-500G
100201-856EA 193.91 USD
100201-856 100205-268
Sodium aluminate ≥99.9% (Al basis) contains ~8% H2O
Sodium aluminate
Formula: NaAlO₂
Melting Pt: 1800 °C
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00801010
CAS Number: 1302-42-7
EINECS: 215-100-1
UN: 2812
ADR: 8,III
Merck Index: 12,08715

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR