Sodium acetate, anhydrous ≥99% ACS

Supplier: STREM CHEMICALS INC MS

Synonyms: Acetic acid sodium salt

93-1105-250G 93-1105-1000G
100200-506EA 99.08 USD
100200-506 100200-034
Sodium acetate, anhydrous ≥99% ACS
Sodium acetate
Formula: H₃CCOONa
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR