Sodium ≥99.8%, ingot
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1078-500G
93-1078-2500G
100197-192EA
311.38
USD
100197-192
100207-212
Sodium ≥99.8%, ingot
Sodium
Formula:
Na MW: 22.99 g/mol Boiling Pt: 892 °C Melting Pt: 97.8 °C Density: 0.97 Storage Temperature: Ambient |
MDL Number:
MFCD00085307 CAS Number: 7440-23-5 EINECS: 231-132-9 UN: 1428 ADR: 4.3,I Merck Index: 12,08710 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...