Sodium sulfate, anhydrous ≥99.999% (Na basis), PURATREM
Supplier: STREM CHEMICALS INC MS
93-1069-10G
93-1069-50G
100201-850EA
147.2
USD
100201-850
100196-980
Sodium sulfate, anhydrous ≥99.999% (Na basis), PURATREM
Sodium sulphate
Formula:
Na₂SO₄ Boiling Pt: 1429 °C (1013 hPa) Melting Pt: 888 °C Density: 2.68 Storage Temperature: Ambient |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 Merck Index: 12,08829 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...