Sodium sulfate, anhydrous ≥99% ACS
Supplier: STREM CHEMICALS INC MS
Synonyms:
Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt
93-1068-250G
93-1068-1000G
93-1068-5000G
100206-902EA
35.38
USD
100206-902
100197-656
100200-312
Sodium sulfate, anhydrous ≥99% ACS
Sodium sulphate
Formula:
Na₂SO₄ Boiling Pt: 1429 °C (1013 hPa) Melting Pt: 888 °C Density: 2.68 Storage Temperature: Ambient |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 Merck Index: 12,08829 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...