Sodium peroxide ≥93% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1050-50G
93-1050-250G
100201-838EA
104.74
USD
100201-838
100206-006
Sodium peroxide ≥93% ACS
Sodium peroxide
Formula:
Na₂O₂ Melting Pt: 460 °C (dec.) Density: 2.805 Storage Temperature: Ambient |
MDL Number:
MFCD00003497 CAS Number: 1313-60-6 EINECS: 215-209-4 UN: 1504 ADR: 5.1,I Merck Index: 13,08728 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...