Boron nitride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-0512-100G 93-0512-25G
100208-486EA 292.98 USD
100208-486 100197-638
Boron nitride ≥99%
Boron nitride
Formula: BN
MW: 24.82 g/mol
Melting Pt: 2527
Density: 2.25
MDL Number: MFCD00011317
CAS Number: 10043-11-5
EINECS: 233-136-6
Merck Index: 13,01336

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR