Boron carbide ≥99%, powder
Supplier: 0000018203
|
Warning
|
93-0509-50G
93-0509-250G
100197-190EA
142.95
USD
100197-190
100207-756
Boron carbide ≥99%, powder
Boron carbide
Formula:
CB₄ Boiling Pt: > 3500 °C Melting Pt: 2350 °C Density: 2.52 |
MDL Number:
MFCD00011520 CAS Number: 12069-32-8 EINECS: 235-111-5 Merck Index: 12,01374 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...