Lithium chloride ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

93-0313-250G 93-0313-1000G
100208-994EA 298.65 USD
100208-994 100197-628
Lithium chloride ≥99% ACS
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Density: 2.068
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR