Ammonium perrhenate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-0249-1G 93-0249-25G 93-0249-5G
100207-750EA 55.2 USD
100207-750 100201-792 100201-794
Ammonium perrhenate ≥99%
Ammonium perrhenate
Formula: H₄NO₄Re
Melting Pt: Dec.
Density: 3.97
MDL Number: MFCD00012377
CAS Number: 13598-65-7
EINECS: 237-075-6
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR