Ammonium sulfate ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
93-0225-2000G
93-0225-500G
100208-984EA
181.17
USD
100208-984
100200-012
Ammonium sulfate ≥99% ACS
Ammonium sulphate
Formula:
(NH₄)₂SO₄ MW: 132.14 g/mol Melting Pt: 235 °C (dec.) Density: 1.769 Storage Temperature: Ambient |
MDL Number:
MFCD00003391 CAS Number: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 13,00559 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...