Ammonium iodide ≥99.999%, PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-0214-25G 93-0214-5G
100210-116EA 437.35 USD
100210-116 100210-118
Ammonium iodide ≥99.999%, PURATREM
Ammonium iodide
Formula: NH₄I
MW: 144.94 g/mol
Melting Pt: 551 (subl.)
Density: 2.514
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7
Merck Index: 13,00531

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR