Ammonium fluoride ≥98% ACS

Supplier: 0000018203
Danger

Synonyms: Fluoroammonium

93-0212-500G 93-0212-100G
100196-414EA 66.52 USD
100196-414 100198-274
Ammonium fluoride ≥98% ACS
Ammonium fluoride
Formula: NH₄F
MW: 37.04 g/mol
Melting Pt: (subl.)
Density: 1.009
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 13,00524

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR