Ammonium chloride ACS

Supplier: STREM CHEMICALS INC MS
Warning

93-0208-500G 93-0208-2000G
100201-788EA 72.18 USD
100201-788 100200-010
Ammonium chloride ACS
Ammonium chloride
Formula: NH₄Cl
Boiling Pt: 520 °C
Melting Pt: 340 °C (subl.)
Density: 1.527
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR