Tetraphenyllead ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

82-2150-1G 82-2150-25G 82-2150-5G
100206-862EA 35.38 USD
100206-862 100201-044 100201-778
Tetraphenyllead ≥97%
Tetraphenyllead
Formula: C₂₄H₂₀Pb
MW: 515.62 g/mol
Boiling Pt: 325 °C/ 230 °C (0.05 mmHg) (dec./ subl.)
Melting Pt: 229…230 °C
Density: 1.530
MDL Number: MFCD00002999
CAS Number: 595-89-1
EINECS: 209-871-3
UN: 3467
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR