Platinum(IV) chloride ≥99.9% (Pt basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Platinum tetrachloride

78-1490-1G 78-1490-5G
100201-752EA 223.63 USD
100201-752 100201-754
Platinum(IV) chloride ≥99.9% (Pt basis)
Platinum (IV) chloride
Formula: Cl₄Pt
Melting Pt: 370 °C (dec.)
Density: 4.303
MDL Number: MFCD00011182
CAS Number: 13454-96-1
EINECS: 236-645-1
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR