Protein Concentrator Powder, G-Biosciences
Supplier: G-Biosciences
786-144 786-144
82021-320EA 180.39 USD
82021-320 CA82021-320
Protein Concentrator Powder, G-Biosciences
Protein Purification Protein Purification Reagents
Concentrator Powder is a high molecular weight polymer which will not migrate across the dialysis membrane
Simply transfer your dilute protein solution to a dialysis bag or dialysis device, such as our patented Tube-O-DIALYZER™ and then cover the membrane with Concentrator Powder. Concentrator Powder rapidly absorbs water from the sample and reduces the sample volume.
Ordering information: Accessories not included.