VWR® UV Crosslinker
Supplier: VWR
VWR®
95-0556-02 95-0556-01
89131-486EA 4918.51 USD
89131-486 89131-484
VWR® UV Crosslinker
UV Crosslinkers
The VWR® Crosslinker’s overhead 254nm UV is used not only for attaching nucleic acids to a membrane, but also for sterilization and decontamination of plates, small tools, etc.
  • Preset and manual controls
  • Flexible and reliable setting controls
  • Large LED display
  • Pull out drawer for easy sample placement
  • Door window blocks UV radiation but still allows observation
  • Five 8-watt 254nm ultraviolet tubes for uniform overhead UV illumination
The Crosslinker assures consistent UV output for many applications including DNA bonding. Features factory presettings that deliver 120,000 microjoules or 5 minutes of exposure; the maximum UV energy setting is 999,900 microjoules per cm2. Microprocessor optimizes the use of energy while ensuring consistent, maximum energy efficiency and output.