Trifluoromethanesulfonic anhydride ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Triflic anhydride
09-7420-25G
09-7420-5G
100209-632EA
171.26
USD
100209-632
100196-022
Trifluoromethanesulfonic anhydride ≥97%
Trifluoromethanesulphonic anhydride
Organo Fluorine
Formula:
C₂F₆O₅S₂ MW: 282.14 g/mol Boiling Pt: 83 Density: 1.677 Storage Temperature: Refrigerator |
MDL Number:
MFCD00000408 CAS Number: 358-23-6 EINECS: 206-616-8 UN: 1760 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...