2,2,2-Trifluoroethanol ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Trifluoro Ethanol, TFE

09-7310-250G 09-7310-50G
100199-746EA 274.58 USD
100199-746 100199-748
2,2,2-Trifluoroethanol ≥99%
2,2,2-Trifluoroethanol

Organo Fluorine

Formula: C₂H₃F₃O
MW: 100.04 g/mol
Boiling Pt: 74.1
Melting Pt: -43.5
Density: 1.373
Flash Pt: 85 °F
Storage Temperature: Ambient
MDL Number: MFCD00004672
CAS Number: 75-89-8
EINECS: 200-913-6
UN: 1992
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR