Trifluoroacetic anhydride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Trifluoracetic anhydrideTFAA

09-7170-50G 09-7170-250G
100198-104EA 113.23 USD
100198-104 100198-102
Trifluoroacetic anhydride ≥98%
Trifluoroacetic anhydride

Organo Fluorine

Formula: C₄F₆O₃
MW: 210.03 g/mol
Boiling Pt: 39…40 °C
Melting Pt: –65 °C
Density: 1.487
MDL Number: MFCD00000416
CAS Number: 407-25-0
EINECS: 206-982-9
UN: 1760
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR