Tantalum ≥99.95%, wire
Supplier: STREM CHEMICALS INC MS
|
Warning
|
73-0075-1MT
73-0075-25MT
73-0075-5MT
100203-270EA
150.03
USD
100203-270
100196-964
100199-258
Tantalum ≥99.95%, wire
Tantalum
Formula:
Ta MW: 180.95 g/mol Boiling Pt: 5427 Melting Pt: 2996 Density: 16.6 |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...