Tantalum ≥99.95%, rods
Supplier: STREM CHEMICALS INC MS
|
Warning
|
73-0070-5CM
73-0070-25CM
100210-830EA
226.46
USD
100210-830
100207-432
Tantalum ≥99.95%, rods
Tantalum
Formula:
Ta MW: 180.95 g/mol Boiling Pt: 5427 Melting Pt: 2996 Density: 16.6 |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...