Hafnium ≥99.7% (Zr-~3%), crystal bar chips

Supplier: STREM CHEMICALS INC MS

72-0055-25G 72-0055-100G
100201-016EA 165.6 USD
100201-016 100196-330
Hafnium ≥99.7% (Zr-~3%), crystal bar chips
Hafnium
Formula: Hf
Boiling Pt: 4602 °C
Melting Pt: 2227 °C
Density: 13,31
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR