Ethylenetetracarbonitrile ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetracyanoethylene

07-1975-25G 07-1975-5G
100209-552EA 593.05 USD
100209-552 100199-330
Ethylenetetracarbonitrile ≥98%
Ethylenetetracarbonitrile
Formula: C₆N₄
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 197…199 °C
Density: 1.35 g/cm³ (25 °C)
MDL Number: MFCD00001850
CAS Number: 670-54-2
EINECS: 211-578-0
UN: 2811
ADR: 6.1,I
Merck Index: 12,09336

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR