Tetrabutylammonium hydroxide 40% water

Supplier: STREM CHEMICALS INC MS
Danger

07-1967-100G 07-1967-25G
100204-480EA 134.46 USD
100204-480 100204-812
Tetrabutylammonium hydroxide 40% water
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48
Melting Pt: 29.85
Density: 1.000
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR