1,10-Phenanthroline, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: o-Phenanthroline

07-1650-25G 07-1650-100G 07-1650-5G
100207-228EA 281.66 USD
100207-228 100208-272 100202-870
1,10-Phenanthroline, anhydrous ≥99%
1,10-Phenanthroline
Formula: C₁₂H₈N₂
MW: 180.21 g/mol
Boiling Pt: 360
Melting Pt: 114…117 °C
MDL Number: MFCD00011678
CAS Number: 66-71-7
EINECS: 200-629-2
UN: 2811
ADR: 6.1 III
Merck Index: 12,07356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5092a8ec81301ed6ed30f75429f9f27784db47a5&vid=106c4325bb7c310de902d3118cf4b31bcee3dc92&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR