Diethylenetriaminepentaacetic acid ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
DTPA
, Idranal V
, Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid
, 2,2',2'',2'''-((((Carboxymethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetic acid
, Diethylenetriaminepentaacetic acid
, N-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid)
, Kestranal® Aza Pa
, Titriplex® V
07-0410-1000G
07-0410-250G
100210-776EA
215.14
USD
100210-776
100195-976
Diethylenetriaminepentaacetic acid ≥97%
Diethylenetriaminepentaacetic acid
Formula:
C₁₄H₂₃N₃O₁₀ MW: 393.35 g/mol Melting Pt: 220 °C (dec.) Density: 1,558 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004289 CAS Number: 67-43-6 EINECS: 200-652-8 UN: 3077 ADR: 9,III Merck Index: 12,07266 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...