Diethylenetriamine ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: 2,2'-Iminodi(ethylamine), DIEN, Diethylenetriamine

07-0400-500G 07-0400-100G
100195-974EA 73.6 USD
100195-974 100204-798
Diethylenetriamine ≥95%
Diethylenetriamine
Formula: C₄H₁₃N₃
MW: 103.17 g/mol
Boiling Pt: 207
Melting Pt: -39
Density: 0.955
Flash Pt: 215 °F
Storage Temperature: Ambient
MDL Number: MFCD00008171
CAS Number: 111-40-0
EINECS: 203-865-4
UN: 2079
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR