Tellurium ≥99.999%, broken ingot
Supplier: STREM CHEMICALS INC MS
|
Danger
|
52-0070-100G
52-0070-25G
52-0070-500G
100208-036EA
744.49
USD
100208-036
100208-936
100208-674
Tellurium ≥99.999%, broken ingot
Tellurium
Formula:
Te Boiling Pt: 989.8 °C Melting Pt: 450 °C Density: 6.24 Storage Temperature: Ambient |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 UN: 3288 ADR: 6.1,III Merck Index: 12,09273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...