Tellurium ≥99.9999%, broken ingot

Supplier: STREM CHEMICALS INC MS
Danger

52-0030-50G 52-0030-10G
100210-008EA 491.14 USD
100210-008 100210-006
Tellurium ≥99.9999%, broken ingot
Tellurium
Formula: Te
Boiling Pt: 989.8 °C
Melting Pt: 450 °C
Density: 6.24
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3288
ADR: 6.1,III
Merck Index: 12,09273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR