Orthoboric acid ≥99.9995%, PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
05-0205-10G
05-0205-50G
100197-288EA
36.8
USD
100197-288
100200-622
Orthoboric acid ≥99.9995%, PURATREM
ortho-Boric acid
Formula:
B(OH)₃ Boiling Pt: 185 °C (1013 hPa) Melting Pt: 171 °C Density: 1.435 Storage Temperature: Ambient |
MDL Number:
MFCD00011337 CAS Number: 10043-35-3 EINECS: 233-139-2 REACH: 01-2119486683-25 Merck Index: 13,01326 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...