Dibutyltin dichloride ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Dichlorodibutylstannane, Di-n-butyltin dichloride, DBTC

50-1130-50G 50-1130-250G 50-1130-1000G
100196-304EA 62.28 USD
100196-304 100209-994 100203-218
Dibutyltin dichloride ≥95%
Dibutyltin dichloride
Formula: C₈H₁₈Cl₂Sn
MW: 303.85 g/mol
Boiling Pt: 275
Melting Pt: 39…41 °C
Density: 1.36
Storage Temperature: Ambient
MDL Number: MFCD00000518
CAS Number: 683-18-1
EINECS: 211-670-0
UN: 3146
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR