Tin ≥99.998%, foil

Supplier: STREM CHEMICALS INC MS

50-0250-50X50MM 50-0250-100X100MM
100204-082EA 107.57 USD
100204-082 100199-242
Tin ≥99.998%, foil
Tin
Formula: Sn
Boiling Pt: 2260 °C
Melting Pt: 231.9 °C
Density: 7.31
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR