Tin ≥99.999%, shot
Supplier: STREM CHEMICALS INC MS
50-0070-5G
50-0070-25G
100204-078EA
52.37
USD
100204-078
100198-212
Tin ≥99.999%, shot
Tin
Formula:
Sn Boiling Pt: 2260 °C Melting Pt: 231.9 °C Density: 7.31 Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...