Silver nitrate ≥99.9995% (Ag basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

47-1325-50G 47-1325-10G
100198-852EA 406.22 USD
100198-852 100204-076
Silver nitrate ≥99.9995% (Ag basis), PURATREM
Formula: AgNO₃
Boiling Pt: 444 °C (dec.)
Melting Pt: 212 °C
Density: 4.352
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR