Palladium ≥99.9%, sponge
Supplier: STREM CHEMICALS INC MS
|
Warning
|
46-2119-1G
46-2119-5G
100211-116EA
213.72
USD
100211-116
100211-118
Palladium ≥99.9%, sponge
Palladium
Formula:
Pd Boiling Pt: 3140 °C Melting Pt: 1552 °C Density: 12.02 Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 Merck Index: 12,07121 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...