Palladium ≥99.95%, rods, Length 20 mm, Diameter 6.35 mm

Supplier: STREM CHEMICALS INC MS
Warning

46-2113-20MM 46-2113-100MM
100209-266EA 1716.86 USD
100209-266 100203-206
Palladium ≥99.95%, rods, Length 20 mm, Diameter 6.35 mm
Palladium
Formula: Pd
Boiling Pt: 3140 °C
Melting Pt: 1552 °C
Density: 12.02
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR