Palladium(II) chloride ≥99.9% (Pd basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Palladium dichloride

46-1850-25G 46-1850-1G 46-1850-5G
100203-204EA 3696.98 USD
100203-204 100208-444 100196-268
Palladium(II) chloride ≥99.9% (Pd basis)
Palladium (II) chloride
Formula: PdCl₂
Melting Pt: 500 °C (dec.)
Density: 4.0
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 13,07058

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR