Niobium(V) oxide ≥99.995% (Nb basis) (50-100ppm Ta), PURATREM

Supplier: STREM CHEMICALS INC MS

Synonyms: Diniobium pentaoxide, Niobium pentoxide

41-5200-100G 41-5200-25G
100197-124EA 379.32 USD
100197-124 100206-608
Niobium(V) oxide ≥99.995% (Nb basis) (50-100ppm Ta), PURATREM
Niobium (V) oxide
Formula: Nb₂O₅
MW: 265.81 g/mol
Melting Pt: 1496
Density: 4.47
MDL Number: MFCD00011128
CAS Number: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR