Niobium ≥99.8%, foil

Supplier: STREM CHEMICALS INC MS
Warning

41-0050-100X50MM 41-0050-100X200MM 41-0050-500X200MM
100198-180EA 209.48 USD
100198-180 100203-124 100203-122
Niobium ≥99.8%, foil
Niobium
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR