Zirconium ≥99.8%, rods
Supplier: STREM CHEMICALS INC MS
40-0080-30CM
40-0080-5CM
100208-888EA
325.54
USD
100208-888
100196-202
Zirconium ≥99.8%, rods
Zirconium
Formula:
Zr MW: 91.22 g/mol Boiling Pt: 3577 °C Melting Pt: 1852 °C Density: 6.506 |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...