Rubidium ≥99.9%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Breakseal ampoule
37-3736-5G
37-3736-1G
100209-338EA
277.42
USD
100209-338
100203-770
Rubidium ≥99.9%
Rubidium
Formula:
Rb Boiling Pt: 688 °C Melting Pt: 38.9 °C Density: 1.532 |
MDL Number:
MFCD00134055 CAS Number: 7440-17-7 EINECS: 231-126-6 UN: 1423 ADR: 4.3,I Merck Index: 12,08439 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...