Phenylselenol ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Benzeneselenol, Selenophenol
34-1600-5G
34-1600-1G
100198-746EA
363.75
USD
100198-746
100204-042
Phenylselenol ≥97%
Phenylselenol
Formula:
C₆H₆Se MW: 157.07 g/mol Boiling Pt: 67…68 °C (12 mmHg) Density: 1,479 Flash Pt: 158 °F |
MDL Number:
MFCD00000001 CAS Number: 645-96-5 EINECS: 211-457-2 UN: 3440 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...