Lithium carbonate ≥99.999% (Li basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
03-0800-100G
03-0800-25G
100207-226EA
339.69
USD
100207-226
100203-884
Lithium carbonate ≥99.999% (Li basis), PURATREM
Lithium carbonate
Formula:
Li₂CO₃ Boiling Pt: 1310 °C (dec.) Melting Pt: 723 °C Density: 2.11 Storage Temperature: Ambient |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 EINECS: 209-062-5 Merck Index: 13,05549 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...