Zinc chloride, anhydrous ≥97% ACS

Supplier: STREM CHEMICALS INC MS
Danger

30-3006-500G 30-3006-100G
100200-922EA 142.95 USD
100200-922 100204-038
Zinc chloride, anhydrous ≥97% ACS
Zinc chloride
Formula: ZnCl₂
Boiling Pt: 732 °C
Melting Pt: 283 °C
Density: 2.91
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR