Zinc ≥99.99%, shot

Supplier: STREM CHEMICALS INC MS

30-0080-1000G 30-0080-250G
100208-018EA 142.95 USD
100208-018 100204-032
Zinc ≥99.99%, shot
Zinc
Formula: Zn
Boiling Pt: 907 °C
Melting Pt: 420 °C
Density: 7.14
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR